Learn More
Medchemexpress LLC 4-Aminohippuric acid-d4 | 1219805-41-0 | 98.0% | 1 MG
SDP
Catalog No. 5000411545
Encompass_Preferred
4-Aminohippuric acid-d4 is a deuterium-labeled version of 4-Aminohippuric acid, which acts as a coordination ligand for various metal ions and a functionalization reagent for nanomaterials. It can form stable complexes with metal ions or function as a reducing agent/stabilizer in nanomaterial synthesis. This compound is useful for constructing highly sensitive electrochemical or colorimetric sensors for detecting and quantifying heavy metal ions in environmental water and biological samples. It may also serve as a biomarker for attention-deficit/hyperactivity disorder (ADHD).
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis through techniques like NMR, GC-MS, or LC-MS.
Catalog No. 50-004-11545 Supplier Medchemexpress LLC Supplier No. HYB1306S1MG
4-Aminohippuric acid-d4 is a deuterium-labeled version of 4-Aminohippuric acid, which acts as a coordination ligand for various metal ions and a functionalization reagent for nanomaterials. It can form stable complexes with metal ions or function as a reducing agent/stabilizer in nanomaterial synthesis. This compound is useful for constructing highly sensitive electrochemical or colorimetric sensors for detecting and quantifying heavy metal ions in environmental water and biological samples. It may also serve as a biomarker for attention-deficit/hyperactivity disorder (ADHD).
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis through techniques like NMR, GC-MS, or LC-MS.