Learn More
Medchemexpress LLC 4-(aminomethyl)-2-methoxyphenol hydrochloride | 7149-10-2 | MFCD00012864 | 99.8% | 189.64 g/mol | C8H12ClNO2 | 25 G
SDP
Catalog No. 5000303521
Encompass_Preferred
Vanillylamine (hydrochloride) is the hydrochloride salt of vanillylamine supplied for research use as a biochemical reagent. It is a crystalline, high-purity intermediate used in organic synthesis and biochemical studies, including the synthesis of capsaicinoids and related vanillyl derivatives.
  • High purity suitable for analytical and synthetic applications (reported ~99.8%).
  • Molecular formula C8H12ClNO2 and molecular weight 189.64 g/mol.
  • Available in multiple small-scale and bulk pack sizes for laboratory use.
  • Useful as a building block for synthesis of capsaicinoids and other vanillyl compounds.
  • Intended for research use only; not for clinical or patient use.
Catalog No. 50-003-03521 Supplier Medchemexpress LLC Supplier No. 25GHYW015211
Vanillylamine (hydrochloride) is the hydrochloride salt of vanillylamine supplied for research use as a biochemical reagent. It is a crystalline, high-purity intermediate used in organic synthesis and biochemical studies, including the synthesis of capsaicinoids and related vanillyl derivatives.
  • High purity suitable for analytical and synthetic applications (reported ~99.8%).
  • Molecular formula C8H12ClNO2 and molecular weight 189.64 g/mol.
  • Available in multiple small-scale and bulk pack sizes for laboratory use.
  • Useful as a building block for synthesis of capsaicinoids and other vanillyl compounds.
  • Intended for research use only; not for clinical or patient use.