Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Aminomethyl)cyclohexanecarboxylic Acid (cis- and trans- mixture) 95.0+%, TCI America™
SDP

Catalog No. A21215G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A21215G Supplier TCI America Supplier No. A21215G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 701-54-2
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.21
MDL Number MFCD00001466,MFCD00064951,MFCD19706018
InChI Key GYDJEQRTZSCIOI-UHFFFAOYSA-N
PubChem CID 5526
IUPAC Name 4-(aminomethyl)cyclohexane-1-carboxylic acid
SMILES NCC1CCC(CC1)C(O)=O

Specifications

Melting Point 233°C
Color White-Yellow
Quantity 5 g
Formula Weight 157.21
Percent Purity ≥95.0% (GC,T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material 4-(Aminomethyl)cyclohexanecarboxylic Acid (cis- and trans- mixture)
RTECSNumber GU8399950
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.