Learn More
Apexbio Technology LLC 4-AMINOMETHYLBENZOIC-ACID-1G
SDP
Catalog No. 5000568836
Encompass_Preferred
4-(Aminomethyl)benzoic acid (CAS No 56-91-7) is a compound primarily studied in the field of biomedical research due to its potential therapeutic properties Originating from synthetic sources it is known to engage in inhibitory mechanisms within specific biological pathways often targeting fibrinolytic activity Its potency is typically observed in the nanomolar to low micromolar range during in vitro assays making it a subject of interest for enzyme inhibition studies The compound is frequently used in various cell models and animal studies particularly for investigating its efficacy and safety as part of drug screening processes Experimental concentrations typically depend on the specific research design allowing for tailored studies based on the intended outcomes and methodologies Its role in modulating biological pathways aids researchers in exploring new therapeutic avenues for diseases where fibrinolysis is a contributing factor
Catalog No. 50-005-68836 Supplier Apexbio Technology LLC Supplier No. C65361000
Add to Cart
Add to Cart
4-(Aminomethyl)benzoic acid (CAS No 56-91-7) is a compound primarily studied in the field of biomedical research due to its potential therapeutic properties Originating from synthetic sources it is known to engage in inhibitory mechanisms within specific biological pathways often targeting fibrinolytic activity Its potency is typically observed in the nanomolar to low micromolar range during in vitro assays making it a subject of interest for enzyme inhibition studies The compound is frequently used in various cell models and animal studies particularly for investigating its efficacy and safety as part of drug screening processes Experimental concentrations typically depend on the specific research design allowing for tailored studies based on the intended outcomes and methodologies Its role in modulating biological pathways aids researchers in exploring new therapeutic avenues for diseases where fibrinolysis is a contributing factor