Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminomorpholine, 95%

Catalog No. AAB2138122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2138122 100 g
AAB2138106 5 g
AAB2138114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2138122 Supplier Thermo Scientific Chemicals Supplier No. B2138122
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 4319-49-7 | C4H10N2O | 102.137 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4319-49-7
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.137
MDL Number MFCD00006174
InChI Key MKQLBNJQQZRQJU-UHFFFAOYSA-N
Synonym 4-aminomorpholine, n-aminomorpholine, 4-morpholinamine, morpholin-4-ylamine, morpholine, 4-amino, 4-morpholinamine 9ci, morpholine-4-ylamine, n-amino morpholine, n-amino-morpholine, 4-amino morpholine
PubChem CID 20315
IUPAC Name morpholin-4-amine
SMILES C1COCCN1N

Specifications

Density 1.059
Boiling Point 168°C
Flash Point 58°C (137°F)
Refractive Index 1.4785
Quantity 100 g
UN Number UN2733
Sensitivity Air sensitive
Formula Weight 102.14
Percent Purity 98%
Chemical Name or Material 4-Aminomorpholine
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H314-H318-H226-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 224-347-4
TSCA No
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.