Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminophenethyl alcohol, 97%

Catalog No. AC400931000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC400931000 100 g Glass bottle
AC400930250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC400931000 Supplier Thermo Scientific Chemicals Supplier No. 400931000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 104-10-9 | C8H11NO | 137.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-10-9
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00007922
InChI Key QXHDYMUPPXAMPQ-UHFFFAOYSA-N
Synonym 2-4-aminophenyl ethanol, 4-aminophenethyl alcohol, p-aminophenethyl alcohol, p-aminophenylethanol, benzeneethanol, 4-amino, 2-p-aminophenyl ethanol, 2-4-aminophenyl ethan-1-ol, 4-2-hydroxyethyl aniline, benzeneethanol, ar-amino, p-2-hydroxyethyl aniline
PubChem CID 66904
IUPAC Name 2-(4-aminophenyl)ethanol
SMILES C1=CC(=CC=C1CCO)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 104.0°C to 108.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula NH2C6H4(CH2)2OH
Quantity 100 g
Beilstein 13,I,242
Solubility Information Solubility in water: slightly soluble.
Formula Weight 137.18
Percent Purity 97%
Physical Form Crystals
Chemical Name or Material 4-Aminophenethyl alcohol, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 203-174-8
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.