Learn More
U.S. Pharmacopeia 4-Aminophenol, 123-30-8, MFCD00007869, 100mg
Catalog No. NC0084364
Encompass
Molecular Formula C6H7NO, Molecular Weight 109.127, Melting Point 356 - 374 °F (180 - 190 °C), Boiling Point 543.2 °F (284 °C) (decomposes), Synonyms: p-Aminophenol, 4-Amino-1-hydroxybenzene
Catalog No. NC0084364 Supplier U.S. Pharmacopeia Supplier No. 1021204
Molecular Formula C6H7NO, Molecular Weight 109.127, Melting Point 356 - 374 °F (180 - 190 °C), Boiling Point 543.2 °F (284 °C) (decomposes), Synonyms: p-Aminophenol, 4-Amino-1-hydroxybenzene