Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminophenol, 97%

Catalog No. ac104270050
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104270050 5 g Glass bottle
AC104272500 250 g Glass bottle
AC104270010 1 kg Glass bottle
AC104270051 5 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC104270050 Supplier Thermo Scientific Chemicals Supplier No. 104270050
Only null left
Add to Cart

CAS: 123-30-8 | C6H7NO | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-30-8
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
InChI Key PLIKAWJENQZMHA-UHFFFAOYSA-N
Synonym p-aminophenol, 4-hydroxyaniline, p-hydroxyaniline, phenol, 4-amino, paranol, 4-aminobenzenol, certinal, citol, azol, fouramine p
PubChem CID 403
ChEBI CHEBI:17602
IUPAC Name 4-aminophenol
SMILES C1=CC(=CC=C1N)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 187.0°C to 191.0°C
Color White to Cream
Boiling Point 284.0°C
Flash Point 189°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H4OH
Quantity 5 g
Beilstein 13,427
Merck Index 15,458
Solubility Information Solubility in water: 15g/L (20°C). Other solubilities: soluble in alcohol, practically insoluble in benzene and chloroform, 9.3% in ethyl methyl ketone (58.5°C), 4.5% in abs.ethanol (0°C)
Formula Weight 109.13
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminophenol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 204-616-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.