Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminophenol (Acetaminophen Related Compound K) (Paracetamol Impurity K) Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019399 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9399 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9399 Supplier MilliporeSigma Supelco Supplier No. PHR11481G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 123-30-8
Flash Point 195°C - Closed Cup
Physical Form Neat
Linear Formula H2 NC6H4 OH
MDL Number MFCD00007869
Melting Point 185°C to 189°C
Synonym 4-Aminophenol; 4-Hydroxyaniline
Type Certified Reference Material
Quantity 1 g
Formula Weight 109.13
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.