Learn More
Medchemexpress LLC 4-aminophenyl β-D-glucopyranoside | 20818-25-1 | MFCD00067365 | 98.9% | 271.27 g/mol | C12H17NO6 | 100 MG
SDP
Catalog No. 5000283817
Encompass_Preferred
4-Aminophenyl β-D-glucopyranoside is a small-molecule biochemical reagent used as a substrate in enzymatic assays and glycobiology research. It is supplied for research use only and typically provided as a solid with high purity suitable for sensitive biochemical applications. Molecular formula C12H17NO6; molecular weight 271.27 g/mol.
  • High purity (≈98.9%) suitable for sensitive biochemical assays.
  • Solid form supplied in common pack sizes including 100 mg.
  • Functions as a substrate for β-glucosidase and other enzymatic studies.
  • Known molecular formula C12H17NO6 and molecular weight 271.27 g/mol for assay calculations.
  • Follow the supplier safety data sheet for handling and storage instructions.
Catalog No. 50-002-83817 Supplier Medchemexpress LLC Supplier No. HYW145490100MG
4-Aminophenyl β-D-glucopyranoside is a small-molecule biochemical reagent used as a substrate in enzymatic assays and glycobiology research. It is supplied for research use only and typically provided as a solid with high purity suitable for sensitive biochemical applications. Molecular formula C12H17NO6; molecular weight 271.27 g/mol.
  • High purity (≈98.9%) suitable for sensitive biochemical assays.
  • Solid form supplied in common pack sizes including 100 mg.
  • Functions as a substrate for β-glucosidase and other enzymatic studies.
  • Known molecular formula C12H17NO6 and molecular weight 271.27 g/mol for assay calculations.
  • Follow the supplier safety data sheet for handling and storage instructions.