Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-aminophenyl β-D-glucopyranoside | 20818-25-1 | MFCD00067365 | 98.9% | 271.27 g/mol | C12H17NO6 | 50 MG
SDP

Catalog No. 5000283767
Encompass_Preferred

4-Aminophenyl β-D-glucopyranoside is a biochemical reagent used in life-science research, commonly employed as a substrate in enzymatic assays, for conjugation or derivatization, and as a synthetic building block. The solid material is characterized by analytic data and safety documentation suitable for laboratory use.

  • High purity (98.9%).
  • Chemical formula C12H17NO6.
  • Molecular weight 271.27 g/mol.
  • CAS number 20818-25-1.
  • Available in mg-scale packages for assay development.
  • Safety data sheets and technical documentation available.

Catalog No. 50-002-83767 Supplier Medchemexpress LLC Supplier No. HYW14549050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Aminophenyl β-D-glucopyranoside is a biochemical reagent used in life-science research, commonly employed as a substrate in enzymatic assays, for conjugation or derivatization, and as a synthetic building block. The solid material is characterized by analytic data and safety documentation suitable for laboratory use.

  • High purity (98.9%).
  • Chemical formula C12H17NO6.
  • Molecular weight 271.27 g/mol.
  • CAS number 20818-25-1.
  • Available in mg-scale packages for assay development.
  • Safety data sheets and technical documentation available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.