Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminophenyl ether, 98%

Catalog No. AC104330250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104330250 25 g Glass bottle
AC104331000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC104330250 Supplier Thermo Scientific Chemicals Supplier No. 104330250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 101-80-4 | C12H12N2O | 200.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-80-4
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.24
MDL Number MFCD00007863
InChI Key HLBLWEWZXPIGSM-UHFFFAOYSA-N
Synonym 4,4'-oxydianiline, 4,4'-diaminodiphenyl ether, 4,4-oxydianiline, 4-aminophenyl ether, 4,4'-oxybisbenzenamine, oxydianiline, 4-4-aminophenoxy aniline, diaminodiphenyl ether, benzenamine, 4,4'-oxybis, p,p'-oxydianiline
PubChem CID 7579
ChEBI CHEBI:34384
IUPAC Name 4-(4-aminophenoxy)aniline
SMILES C1=CC(=CC=C1N)OC2=CC=C(C=C2)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 189°C to 193°C
Boiling Point 190°C (0.1 mmHg)
Flash Point 218°C
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (H2NC6H4)2O
Quantity 25 g
Beilstein 13, 441
Solubility Information Solubility in water: negligible
Formula Weight 200.24
Percent Purity 98%
Chemical Name or Material 4-Aminophenyl ether
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause cancer.
May cause genetic defects.
Suspected of damaging fertility.
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-977-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.