Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminophenyl ether, 98%

Catalog No. AC104335000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC104335000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC104335000 Supplier Thermo Scientific Chemicals Supplier No. 104335000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 101-80-4 | C12H12N2O | 200.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-80-4
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.24
InChI Key HLBLWEWZXPIGSM-UHFFFAOYSA-N
Synonym 4,4'-oxydianiline, 4,4'-diaminodiphenyl ether, 4,4-oxydianiline, 4-aminophenyl ether, 4,4'-oxybisbenzenamine, oxydianiline, 4-4-aminophenoxy aniline, diaminodiphenyl ether, benzenamine, 4,4'-oxybis, p,p'-oxydianiline
PubChem CID 7579
ChEBI CHEBI:34384
IUPAC Name 4-(4-aminophenoxy)aniline
SMILES C1=CC(=CC=C1N)OC2=CC=C(C=C2)N

Specifications

Melting Point 189.0°C to 193.0°C
Boiling Point 190.0°C (0.1 mmHg)
Flash Point 218°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula (H2NC6H4)2O
Quantity 500 g
Beilstein 13,441
Solubility Information Solubility in water: negligible.
Formula Weight 200.24
Percent Purity 98%
Chemical Name or Material 4-Aminophenyl ether
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement May cause cancer. May cause genetic defects. Suspected of damaging fertility. Toxic if inhaled. Toxic in contact with skin. Toxic if swallowed. Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF exposed or concerned: Get medical advice/attention. Avoid release to the environment.
EINECSNumber 202-977-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.