Learn More
Cayman Chemical 4-Aminophenyl Phosphate (sodium salt hydrate), Purity: ≥95%, 100 mg
SDP
Catalog No. 502967378
Encompass_Preferred
A substrate for alkaline phosphatase that can be used in enzymatic reactions to quantify ALP activity
Catalog No. 50-296-7378 Supplier Cayman Chemical Supplier No. 17293100 MG
A substrate for alkaline phosphatase that can be used in enzymatic reactions to quantify ALP activity