Learn More
Cayman Chemical 4-Aminophenyl Phosphate sodi
SDP
Catalog No. 502965466
Encompass_Preferred
A substrate for alkaline phosphatase that can be used in enzymatic reactions to quantify ALP activity
Catalog No. 50-296-5466 Supplier Cayman Chemical Supplier No. 17293500 MG
A substrate for alkaline phosphatase that can be used in enzymatic reactions to quantify ALP activity