Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminophenylarsonic Acid 98.0+%, TCI America™
SDP

Catalog No. A0530500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0530500G 500 g
A053025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0530500G Supplier TCI America Supplier No. A0530500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 98-50-0
Molecular Formula C6H8AsNO3
Molecular Weight (g/mol) 217.056
MDL Number MFCD00007819
InChI Key XKNKHVGWJDPIRJ-UHFFFAOYSA-N
Synonym p-arsanilic acid, arsanilic acid, 4-arsanilic acid, 4-aminophenyl arsonic acid, 4-aminobenzenearsonic acid, atoxylic acid, p-aminobenzenearsonic acid, aminophenylarsine acid, p-anilinearsonic acid, pro-gen
PubChem CID 7389
ChEBI CHEBI:49477
IUPAC Name (4-aminophenyl)arsonic acid
SMILES C1=CC(=CC=C1N)[As](=O)(O)O

Specifications

Quantity 500 g
UN Number 3465
Formula Weight 217.06
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminophenylarsonic Acid
RTECSNumber CF7875000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.