Learn More
AdipoGen 4-Aminophenylphosphate . sodium salt (50 mg)
SDP
Supplier:  AdipoGen AGCR13586M050
Encompass_Preferred
Chemical. CAS: 108084-47-5 | 52331-30-3 (free base). Formula: C6H7NO4P . Na. MW: 188.1 . 23.0. Substrate for the electrochemical measurement of alkaline phosphatase.
Catalog No. 50-205-3952