Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminopyrazolo[3,4-d]pyrimidine 97.0+%, TCI America™
SDP

Catalog No. A1041100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1041100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1041100MG Supplier TCI America Supplier No. A1041100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2380-63-4
Molecular Formula C5H5N5
Molecular Weight (g/mol) 135.13
MDL Number MFCD00005688
InChI Key LHCPRYRLDOSKHK-UHFFFAOYSA-N
Synonym 1h-pyrazolo 3,4-d pyrimidin-4-amine, 4-aminopyrazolo 3,4-d pyrimidine, 4-amino-1h-pyrazolo 3,4-d pyrimidine, pyrazoloadenine, 1h-pyrazolo 3,4-d pyrimidin-4-ylamine, 4-aminopyrazolopyrimidine, 4 app, 4-app, adenine antimetabolite
PubChem CID 75420
IUPAC Name 1H-pyrazolo[3,4-d]pyrimidin-4-amine
SMILES NC1=C2C=NNC2=NC=N1

Specifications

Color White-Yellow
Quantity 100 mg
UN Number 2811
Formula Weight 135.13
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminopyrazolo[3,4-d]pyrimidine
RTECSNumber UR0717000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.