Learn More
Selleck Chemical LLC 4-Aminopyridine-25mg
SDP
Supplier:  Selleck Chemical LLC S502825MG
Encompass_Preferred
4-Aminopyridine (4-AP, Fampridine, Dalfampridine) is a potent and non-selective inhibitor of voltage gated potassium channels (Kv) with IC50 values of 170 μM and 230 μM for Kv1.1 and Kv1.2 in CHO cells, respectively.
Catalog No. 50-238-9853