Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminoquinoline, 95+%, Thermo Scientific™

Catalog No. AC459760010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459760010 1 g
AC459762500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459760010 Supplier Thermo Scientific Chemicals Supplier No. 459760010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 578-68-7
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.17
InChI Key FQYRLEXKXQRZDH-UHFFFAOYSA-N
Synonym 4-aminoquinoline, 4-quinolinamine, 4-aminoguinoline, quinolin-4-ylamine, 4-quinolylamine, unii-gte5p5l97n, ccris 1679, gte5p5l97n, 4-quinolinylamine, 4-amino quinoline
PubChem CID 68476
IUPAC Name quinolin-4-amine
SMILES C1=CC=C2C(=C1)C(=CC=N2)N

Specifications

Boiling Point 332.0°C to 337.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: slightly soluble.
Formula Weight 144.17
Percent Purity 95+%
Chemical Name or Material 4-Aminoquinoline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.