Learn More
Cayman Chemical 4-AmInosalIcylIc AcId 250g
SDP
Catalog No. 502948291
Encompass_Preferred
A PABA antimetabolite with antibacterial activity; active against streptomycin-sensitive and -resistant strains of M. tuberculosis (MICs = 0.78 and 0.39 UG/ml, respectively); an alternative substrate for FolP1 and misincorporation into the folate pathway leads to accumulation of several folate-dependent metabolites including serine, homocysteine, dUMP, and AICAR, markers of folate pathway inhibition, in a concentration-dependent manner; reverses manganese-induced increases in rat hippocampal levels of NLRP3, cleaved caspase-1, and phosphorylated p65 at 300 mg/kg; a synthetic building block
Catalog No. 50-294-8291 Supplier Cayman Chemical Supplier No. 33449250 G
Add to Cart
Add to Cart
A PABA antimetabolite with antibacterial activity; active against streptomycin-sensitive and -resistant strains of M. tuberculosis (MICs = 0.78 and 0.39 UG/ml, respectively); an alternative substrate for FolP1 and misincorporation into the folate pathway leads to accumulation of several folate-dependent metabolites including serine, homocysteine, dUMP, and AICAR, markers of folate pathway inhibition, in a concentration-dependent manner; reverses manganese-induced increases in rat hippocampal levels of NLRP3, cleaved caspase-1, and phosphorylated p65 at 300 mg/kg; a synthetic building block