Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminosulfonyl-1-hydroxy-2-naphthoic Acid 97%, Thermo Scientific™

Catalog No. AC162460100
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC162460100 10 g
AC162460050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC162460100 Supplier Thermo Scientific Chemicals Supplier No. 162460100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64415-15-2
Molecular Formula C11H9NO5S
Molecular Weight (g/mol) 267.255
InChI Key OTDVKOVLSPJORY-UHFFFAOYSA-N
Synonym 4-aminosulphonyl-1-hydroxy-2-naphthoic acid, 1-hydroxy-4-sulfamoyl-2-naphthoic acid, 2-naphthalenecarboxylic acid, 4-aminosulfonyl-1-hydroxy, 4-aminosulfonyl-1-hydroxy-2-naphthoic acid, ncistruc1_001970, ncistruc2_000773, 4-aminosulfonyl-1-hydroxy-2-naphthoic-acid, 4-aminosulfonyl-1-hydroxy-2-naphthalenecarboxylic acid, 1-hydroxy-4-hydroxysulfonimidoyl-2-naphthoic acid #
PubChem CID 97423
IUPAC Name 1-hydroxy-4-sulfamoylnaphthalene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=CC(=C2O)C(=O)O)S(=O)(=O)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown
Infrared Spectrum Authentic
Linear Formula H2NSO2C10H5(OH)CO2H
Quantity 10 g
Formula Weight 267.26
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material 4-Aminosulfonyl-1-hydroxy-2-naphthoic acid, 97%
EINECSNumber 264-882-0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.