Learn More
Medchemexpress LLC 4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole | 91366-65-3 | 98.8% | 25 MG
SDP
Catalog No. 5000413552
Encompass_Preferred
4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole (ABD-F) is a fluorescent reagent used for the sensitive and specific detection of thiols. It has an excitation/emission (Ex/Em) of 380/510 nm. It exhibits low fluorescence background and good stability of fluorophor, making it suitable for various research applications.
  • Sensitive and specific detection of thiols.
  • Low fluorescence background.
  • Good stability of fluorophor.
  • Useful for research purposes.
Catalog No. 50-004-13552 Supplier Medchemexpress LLC Supplier No. 25MGHYD0102
4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole (ABD-F) is a fluorescent reagent used for the sensitive and specific detection of thiols. It has an excitation/emission (Ex/Em) of 380/510 nm. It exhibits low fluorescence background and good stability of fluorophor, making it suitable for various research applications.
  • Sensitive and specific detection of thiols.
  • Low fluorescence background.
  • Good stability of fluorophor.
  • Useful for research purposes.