Learn More
Medchemexpress LLC 4-(aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole | 91366-65-3 | MFCD00036687 | 98.8% | 217.18 | C6H4FN3O3S | 10 MG
SDP
Catalog No. 5000296809
Encompass_Preferred
4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole is a fluorescent reagent for the sensitive and specific detection of thiols. It offers low background fluorescence and a stable fluorophore, making it suitable for thiol derivatization and fluorescence-based analytical applications. The compound is typically supplied as a yellow to green solid and should be protected from light during storage.
  • Provides sensitive and specific fluorescence-based thiol detection.
  • Low fluorescence background for improved signal-to-noise.
  • Stable fluorophore under recommended storage conditions.
  • Supplied in small, lab-scale quantities for analytical use.
  • Yellow to green solid, easy to handle and store.
Catalog No. 50-002-96809 Supplier Medchemexpress LLC Supplier No. HYD010210MG
4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole is a fluorescent reagent for the sensitive and specific detection of thiols. It offers low background fluorescence and a stable fluorophore, making it suitable for thiol derivatization and fluorescence-based analytical applications. The compound is typically supplied as a yellow to green solid and should be protected from light during storage.
  • Provides sensitive and specific fluorescence-based thiol detection.
  • Low fluorescence background for improved signal-to-noise.
  • Stable fluorophore under recommended storage conditions.
  • Supplied in small, lab-scale quantities for analytical use.
  • Yellow to green solid, easy to handle and store.