Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminothiophenol, 96%

Catalog No. p-58971
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC104680050 5 g
AC104680250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC104680050 Supplier Thermo Scientific Chemicals Supplier No. 104680050
Only null left
Add to Cart

CAS: 1193-02-8 | C6H6NS | 124.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-02-8
Molecular Formula C6H6NS
Molecular Weight (g/mol) 124.18
MDL Number MFCD00007880
InChI Key WCDSVWRUXWCYFN-UHFFFAOYSA-M
Synonym 4-aminothiophenol, 4-mercaptoaniline, p-aminothiophenol, p-aminobenzenethiol, 4-amino thiophenol, benzenethiol, 4-amino, p-mercaptoaniline, benzenethiol, p-amino, p-aminophenylmercaptan, 4-aminophenyl mercaptan
PubChem CID 14510
IUPAC Name 4-aminobenzenethiol
SMILES NC1=CC=C([S-])C=C1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 37.0°C to 43.0°C
Color White to Brown
Boiling Point 140.0°C to 145.0°C (16.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (Argentometry)
Packaging Glass bottle
Linear Formula H2NC6H4SH
Quantity 5 g
Beilstein 13, 533
Formula Weight 125.19
Percent Purity 96%
Physical Form Crystalline Mass, Crystals or Flakes
Chemical Name or Material 4-Aminothiophenol, 96%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 214-763-4
RTECSNumber DC0602500

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.