Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminotoluene-3-sulfonic Acid, MP Biomedicals

Catalog No. ICN213155 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN213155 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN213155 Supplier MP Biomedicals, Inc Supplier No. 0521315580

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 88-44-8
Molecular Formula C7H9NO3S
Molecular Weight (g/mol) 187.21
MDL Number MFCD00007908
InChI Key LTPSRQRIPCVMKQ-UHFFFAOYSA-N
Synonym 4-aminotoluene-3-sulfonic acid, ptmsptmsa, ptms, ptmsa, benzenesulfonic acid, 2-amino-5-methyl, 6-amino-m-toluenesulfonic acid, red 4b acid, p-toluidine-2-sulfonic acid, p-toluidine-m-sulfonic acid, m-toluenesulfonic acid, 6-amino
PubChem CID 6934
IUPAC Name 2-amino-5-methylbenzene-1-sulfonic acid
SMILES CC1=CC=C(N)C(=C1)S(O)(=O)=O

Specifications

Quantity 100 g
Formula Weight 187.22
Chemical Name or Material 4-Aminotoluene-3-sulfonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.