Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amylcyclohexanol (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. A19265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19265G 5 g
A192625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19265G Supplier TCI America Supplier No. A19265G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 54410-90-1
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.296
InChI Key VHWGPISIUNUREA-UHFFFAOYSA-N
Synonym 4-Pentylcyclohexanol
PubChem CID 41076
IUPAC Name 4-pentylcyclohexan-1-ol
SMILES CCCCCC1CCC(CC1)O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 170.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Amylcyclohexanol (cis- and trans- mixture)
RTECSNumber GW0680000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.