Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-(Amyloxy)benzylidene-4-cyanoaniline 97.0+%, TCI America™
SDP

Catalog No. B02531G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B02531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B02531G Supplier TCI America Supplier No. B02531G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 37075-25-5
Molecular Formula C19H20N2O
Molecular Weight (g/mol) 292.382
MDL Number MFCD00059588
InChI Key IUWUDHSOMQRZCV-UHFFFAOYSA-N
Synonym 4′C-(Pentyloxy)benzylidene-4-cyanoaniline, 4-[[4-(Amyloxy)benzylidene]amino]benzonitrile, 4-[[4-(Pentyloxy)benzylidene]amino]benzonitrile
PubChem CID 23369117
IUPAC Name 4-[(4-pentoxyphenyl)methylideneamino]benzonitrile
SMILES CCCCCOC1=CC=C(C=C1)C=NC2=CC=C(C=C2)C#N

Specifications

Color Yellow
Quantity 1 g
Formula Weight 292.38
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-(Amyloxy)benzylidene-4-cyanoaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.