Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amyloxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. A070825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A070825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A070825G Supplier TCI America Supplier No. A070825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15872-41-0
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD00013990
InChI Key OZPPUPJQRJYTNY-UHFFFAOYSA-N
Synonym 4-pentyloxybenzoic acid, 4-pentyloxy benzoic acid, 4-n-pentyloxybenzoic acid, benzoic acid, 4-pentyloxy, p-pentyloxybenzoic acid, 4-n-amyloxybenzoic acid, p-pentoxybenzoic acid, p-n-pentyloxybenzoic acid, 4-amyloxybenzoic acid, p-pentyloxy benzoic acid
PubChem CID 85153
IUPAC Name 4-pentoxybenzoic acid
SMILES CCCCCOC1=CC=C(C=C1)C(=O)O

Specifications

Color White
Quantity 25 g
Formula Weight 208.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amyloxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.