Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amyloxyphenol 97.0+%, TCI America™
SDP

Catalog No. A07285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A07285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A07285G Supplier TCI America Supplier No. A07285G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18979-53-8
Molecular Formula C11H16O2
Molecular Weight (g/mol) 180.25
MDL Number MFCD00044283
InChI Key JCLFHZLOKITRCE-UHFFFAOYSA-N
Synonym Hydroquinone Monoamyl Ether, Hydroquinone Monopentyl Ether, 4-Pentyloxyphenol
PubChem CID 29353
IUPAC Name 4-(pentyloxy)phenol
SMILES CCCCCOC1=CC=C(O)C=C1

Specifications

Melting Point 48°C
Color Red-White
Quantity 5 g
Formula Weight 180.25
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amyloxyphenol
EINECSNumber (3)-3539
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.