Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Anilino-1-methylpiperidine, 98%, Thermo Scientific™

Catalog No. AAH2727614
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2727614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2727614 Supplier Thermo Scientific Chemicals Supplier No. H2727614

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22261-94-5
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD06408133
InChI Key GXJSWUKLJQHQFZ-UHFFFAOYSA-N
PubChem CID 89642
IUPAC Name 1-methyl-N-phenylpiperidin-4-amine
SMILES CN1CCC(CC1)NC2=CC=CC=C2

Specifications

Melting Point 82°C to 86°C
Quantity 25 g
Synonym 4-anilino-1-methylpiperidine, n-1-methylpiperidin-4-yl aniline, 4-piperidinamine,1-methyl-n-phenyl, 1-methyl 4-piperidyl phenylamine, acmc-20akdg, 4-piperidinamine, 1-methyl-n-phenyl, 4-anilino-1-methylpiperidin, 4-anilino-1-methyl-piperidine, n-1-methyl-4-piperidinyl aniline, 1-methyl-n-phenyl-4-piperidinamine
Formula Weight 190.28
Percent Purity 98%
Chemical Name or Material 4-Anilino-1-methylpiperidine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 244-882-7
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.