Learn More
Cayman Chemical 4-APBhydrochlorIde 10mg
SDP
Catalog No. 502954137
Encompass_Preferred
An analytical reference standard categorized as a benzofuran; a potential metabolite of 4-MAPB and 4-EAPB based on the published metabolism of 5-MAPB; intended for research and forensic applications
Catalog No. 50-295-4137 Supplier Cayman Chemical Supplier No. 1428110 MG
An analytical reference standard categorized as a benzofuran; a potential metabolite of 4-MAPB and 4-EAPB based on the published metabolism of 5-MAPB; intended for research and forensic applications