Learn More
Cayman Chemical 4-APDBhydrochlorIde 10mg
SDP
Catalog No. 502893104
Encompass_Preferred
An analytical reference standard categorized as a benzofuran; intended for research and forensic applications
Catalog No. 50-289-3104 Supplier Cayman Chemical Supplier No. 900189110 MG
An analytical reference standard categorized as a benzofuran; intended for research and forensic applications