Learn More
Medchemexpress LLC 4-Arm-PEG-Acrylate | 95.0% | 10000 (Average) | 100 MG
SDP
Catalog No. 5000386793
Encompass_Preferred
4-Arm-PEG-AC (MW 10000) is a polymer consisting of a four-arm PEG core structure with acrylate groups attached to the ends of each PEG chain by chemical reaction. It is for research use only and not sold to patients.
  • Used for drug delivery.
  • For research use only.
Catalog No. 50-003-86793 Supplier Medchemexpress LLC Supplier No. HY174968B100MG
Add to Cart
Add to Cart
4-Arm-PEG-AC (MW 10000) is a polymer consisting of a four-arm PEG core structure with acrylate groups attached to the ends of each PEG chain by chemical reaction. It is for research use only and not sold to patients.
  • Used for drug delivery.
  • For research use only.