Learn More
Creative Pegworks 4-Arm PEG-MAL, MW 2k - 10g
SDP
Catalog No. NC3345727
Encompass
4-Arm PEG-MAL is a multiarmed PEG derivative with maleimide at each terminal of the four arms connected to one pentaerythritol core. Maleimide selectively reacts with free thiol, SH, sulfhydryl, or mercapto group via Michael addition to form a stable thioether bond. PEG-MAL useful for site specific protein and peptide modification. Hydroquinone MEHQ at ~ 200 ppm may be present as inhibitors to prevent self polymerization and partial crosslinking. MEHQ may be removed per customer request.
Catalog No. NC3345727 Supplier Creative Pegworks Supplier No. PSB45010G
4-Arm PEG-MAL is a multiarmed PEG derivative with maleimide at each terminal of the four arms connected to one pentaerythritol core. Maleimide selectively reacts with free thiol, SH, sulfhydryl, or mercapto group via Michael addition to form a stable thioether bond. PEG-MAL useful for site specific protein and peptide modification. Hydroquinone MEHQ at ~ 200 ppm may be present as inhibitors to prevent self polymerization and partial crosslinking. MEHQ may be removed per customer request.