Learn More
Creative Pegworks 4-Arm PEG-OH MW 10k - 10g
SDP
Catalog No. NC2753534
Encompass
4-Arm PEG-OH is a multi-armed polyethylene glycol with hydroxyl groups at each terminal of the four arms. Pentaerythritol core is used in 4-arm PEG raw materials and derivatives. The indicated molecular weight of multi-arm PEG is the sum of the molecular weights of all the arms. 4-Arm PEG-OH can be used to prepare functional PEG reagents or in sol-gel chemistry to make silica-related hydrogel.
Catalog No. NC2753534 Supplier Creative Pegworks Supplier No. PJK40110G
4-Arm PEG-OH is a multi-armed polyethylene glycol with hydroxyl groups at each terminal of the four arms. Pentaerythritol core is used in 4-arm PEG raw materials and derivatives. The indicated molecular weight of multi-arm PEG is the sum of the molecular weights of all the arms. 4-Arm PEG-OH can be used to prepare functional PEG reagents or in sol-gel chemistry to make silica-related hydrogel.