Learn More
Medchemexpress LLC 4-Arm-PEG-OH (MW 20000) | 98.0% | 20000 | 5 MG
SDP
Catalog No. 5000386775
Encompass_Preferred
4-Arm-PEG-OH (MW 20000) is a PEG derivative with a four-arm structure and a hydroxyl (OH) functional group. It reacts with functional groups on biomolecules (such as proteins, peptides, antibodies, etc.) through hydroxyl groups and can be used for drug delivery.
  • PEG derivative with a four-arm structure
  • Hydroxyl (OH) functional group
  • Reacts with functional groups on biomolecules
  • Suitable for drug delivery
Catalog No. 50-003-86775 Supplier Medchemexpress LLC Supplier No. HY174967C5MG
4-Arm-PEG-OH (MW 20000) is a PEG derivative with a four-arm structure and a hydroxyl (OH) functional group. It reacts with functional groups on biomolecules (such as proteins, peptides, antibodies, etc.) through hydroxyl groups and can be used for drug delivery.
  • PEG derivative with a four-arm structure
  • Hydroxyl (OH) functional group
  • Reacts with functional groups on biomolecules
  • Suitable for drug delivery