Learn More
Medchemexpress LLC 4-Arm-PEG-OH (MW 20000) | 98.0% | 20000 (Average) | 100 MG
SDP
Catalog No. 5000387023
Encompass_Preferred
4-Arm-PEG-OH (MW 20000) is a PEG derivative characterized by a four-arm structure and a hydroxyl (OH) functional group. This compound is designed to react with functional groups found on various biomolecules, such as proteins, peptides, and antibodies, through its hydroxyl groups. It is primarily utilized in drug delivery applications.
  • Four-arm structure
  • Hydroxyl (OH) functional group
  • Reacts with biomolecules (proteins, peptides, antibodies)
  • Used for drug delivery
  • Solid, white to off-white appearance
  • Soluble in ethanol (40 mg/mL with sonication)
  • Long-term stability in powder and solvent forms
Catalog No. 50-003-87023 Supplier Medchemexpress LLC Supplier No. HY174967C100MG
Add to Cart
Add to Cart
4-Arm-PEG-OH (MW 20000) is a PEG derivative characterized by a four-arm structure and a hydroxyl (OH) functional group. This compound is designed to react with functional groups found on various biomolecules, such as proteins, peptides, and antibodies, through its hydroxyl groups. It is primarily utilized in drug delivery applications.
  • Four-arm structure
  • Hydroxyl (OH) functional group
  • Reacts with biomolecules (proteins, peptides, antibodies)
  • Used for drug delivery
  • Solid, white to off-white appearance
  • Soluble in ethanol (40 mg/mL with sonication)
  • Long-term stability in powder and solvent forms