Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Azaphthalide, 98%, Thermo Scientific Chemicals

Catalog No. AC458450010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458450010 1 g
AC458450050 5 g
AC458450250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC458450010 Supplier Thermo Scientific Chemicals Supplier No. 458450010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5657-51-2
Molecular Formula C7H5NO2
Molecular Weight (g/mol) 135.12
InChI Key YSJHADWSLVFGGT-UHFFFAOYSA-N
Synonym furo 3,4-b pyridin-5 7h-one, 4-azaphthalide, 7h-furo 3,4-b pyridin-5-one, ksc269g5r, 5h,7h-furo 3,4-b pyridin-5-one
PubChem CID 819439
IUPAC Name 7H-furo[3,4-b]pyridin-5-one
SMILES C1C2=C(C=CC=N2)C(=O)O1

Specifications

Melting Point 143.0°C to 144.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 135.12
Percent Purity 98%
Chemical Name or Material 4-Azaphthalide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.