Learn More
Medchemexpress LLC 4-azido-2,2,6,6-tetramethyl-1-piperidinyloxy | 63697-61-0 | MFCD16659950 | 99.4% | 197.26 g/mol | C9H17N4O | 10 MG
SDP
Catalog No. 5000287663
Encompass_Preferred
N3-TEMPO is an azide-functionalized TEMPO derivative used to introduce a nitroxide spin label via click chemistry. It participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC), enabling efficient conjugation and labeling of biomolecules. The compound is supplied as a high-purity solid for research applications.
  • Azide-functionalized TEMPO for click chemistry.
  • Reacts in CuAAC and SPAAC for efficient conjugation.
  • Introduces a stable nitroxide spin label for EPR and probing studies.
  • High purity suitable for research-grade applications.
  • Solid form for convenient storage and handling.
  • Useful for labeling proteins, nucleic acids, lipids, and small molecules.
Catalog No. 50-002-87663 Supplier Medchemexpress LLC Supplier No. HY15184710MG
N3-TEMPO is an azide-functionalized TEMPO derivative used to introduce a nitroxide spin label via click chemistry. It participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC), enabling efficient conjugation and labeling of biomolecules. The compound is supplied as a high-purity solid for research applications.
  • Azide-functionalized TEMPO for click chemistry.
  • Reacts in CuAAC and SPAAC for efficient conjugation.
  • Introduces a stable nitroxide spin label for EPR and probing studies.
  • High purity suitable for research-grade applications.
  • Solid form for convenient storage and handling.
  • Useful for labeling proteins, nucleic acids, lipids, and small molecules.