Learn More
Medchemexpress LLC HY-16714 500mg Medchemexpress, 4-Azido-L-phenylalanine p-Azidophenylalanine;p-Azido-L-phenylalanine CAS:33173-53-4 Purity:98%
SDP
Catalog No. 501638557
Encompass_Preferred
Medchemexpress,HY-16714 500mg 4-Azido-L-phenylalanine p-Azidophenylalanine;p-Azido-L-phenylalanine CAS:33173-53-4 Formula:C9H10N4O2 4-Azido-L-phenylalanine is an unnatural amino acid, which is used as an effective vibrational reporter of local protein environments. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8557 Supplier Medchemexpress LLC Supplier No. HY16714500MG
Medchemexpress,HY-16714 500mg 4-Azido-L-phenylalanine p-Azidophenylalanine;p-Azido-L-phenylalanine CAS:33173-53-4 Formula:C9H10N4O2 4-Azido-L-phenylalanine is an unnatural amino acid, which is used as an effective vibrational reporter of local protein environments. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.