Learn More
Apexbio Technology LLC 4-Azido-L-phenylalanine 33173-53-4 200mg
SDP
Catalog No. 5000093522
Encompass_Preferred
4-Azido-L-phenylalanine is a noncanonical amino acid containing an azide functional group 4-Azido-L-phenylalanine provides an accessible reactive site suitable for chemoselective click conjugation strategies including copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC) using cyclooctynes such as DBCO and BCN Based on these properties 4-Azido-L-phenylalanine holds research potential in site-specific protein labeling chemical modification and studies of protein-protein interactions cellular protein dynamics and protein functional mechanisms
Catalog No. 50-000-93522 Supplier Apexbio Technology LLC Supplier No. A7094200
4-Azido-L-phenylalanine is a noncanonical amino acid containing an azide functional group 4-Azido-L-phenylalanine provides an accessible reactive site suitable for chemoselective click conjugation strategies including copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC) using cyclooctynes such as DBCO and BCN Based on these properties 4-Azido-L-phenylalanine holds research potential in site-specific protein labeling chemical modification and studies of protein-protein interactions cellular protein dynamics and protein functional mechanisms