Learn More
TARGETMOL CHEMICALS INC 4-AZIDO-L-PHENYLALANINE 500MG
SDP
Catalog No. 502581315
Encompass_Preferred
Also available in 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 4-Azido-L-phenylalanine hydrochloride (p-Azido-L-phenylalanine hydrochloride) is an unnatural amino acid used as an effective vibrational reporter of local protein environments. purity: 99%
Catalog No. 50-258-1315 Supplier TARGETMOL CHEMICALS INC Supplier No. T14038L500MG
Also available in 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 4-Azido-L-phenylalanine hydrochloride (p-Azido-L-phenylalanine hydrochloride) is an unnatural amino acid used as an effective vibrational reporter of local protein environments. purity: 99%