Learn More
Cayman Chemical 4-AzIdo-L-phenylalnIn 250mg
SDP
Catalog No. 502944250
Encompass_Preferred
A derivative of the essential amino acid L-phenylalanine and a click chemistry reagent; has been used as a photocrosslinking agent and in spin labeling for the assessment of protein-protein interactions in vitro and in vivo
Catalog No. 50-294-4250 Supplier Cayman Chemical Supplier No. 41697250 MG
A derivative of the essential amino acid L-phenylalanine and a click chemistry reagent; has been used as a photocrosslinking agent and in spin labeling for the assessment of protein-protein interactions in vitro and in vivo