Learn More
Cayman Chemical 4-Azido-L-phenylalanine, Purity: ≥98%, CAS: 33173-53-4, 500 mg
SDP
Catalog No. 502943431
Encompass_Preferred
A derivative of the essential amino acid L-phenylalanine and a click chemistry reagent; has been used as a photocrosslinking agent and in spin labeling for the assessment of protein-protein interactions in vitro and in vivo
Catalog No. 50-294-3431 Supplier Cayman Chemical Supplier No. 41697500 MG
A derivative of the essential amino acid L-phenylalanine and a click chemistry reagent; has been used as a photocrosslinking agent and in spin labeling for the assessment of protein-protein interactions in vitro and in vivo