Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Azidoaniline hydrochloride

Catalog No. 359556250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-955-6250MG 250 mg
35-955-61G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 35-955-6250MG Supplier Sigma Aldrich Supplier No. 359556250MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Azidoaniline hydrochloride

Specifications

CAS 91159-79-4
Melting Point 165°C (lit.)
Molecular Formula C6H6N4 -+ HCl
Linear Formula N3C6H4NH2 -+ HCl
MDL Number MFCD00043274
Quantity 250 mg
Synonym 4-Aminophenyl azide hydrochloride
Molecular Weight (g/mol) 170.6
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.