Learn More
Description
- Diphenyl selenide is an organoselenium compound
- Its standard enthalpies of combustion, formation and the mean bond-dissociation energy have been calculated
- Its potential as a neutral carrier to develop silver-selective membrane electrode has been investigated
- Diphenyl selenide undergoes oxidation to form selenoxides on treating with n-bromosuccinimide followed by alkaline hydrolysis.used for anti-selective michael addition of thiols and their analogs to nitro-olefins
Specifications
Specifications
| Molecular Formula | C7H7N5O |
| Linear Formula | C7H7N5O |
| MDL Number | MFCD00083160 |
| Quantity | 10 g |
| Molecular Weight (g/mol) | 177.16 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.