Learn More
Medchemexpress LLC 4-Azidobutylamine | 88192-20-5 | 98.0% | 114.15 | 250 MG
SDP
Catalog No. 5000347596
Encompass_Preferred
4-Azidobutylamine is a PROTAC linker, characterized by its alkyl chain composition. It is utilized in the synthesis of PROTACs, which are molecules containing two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for a target protein. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins. Additionally, it is a click chemistry reagent containing an Azide group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules, or strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PROTAC linker
  • Click chemistry reagent
  • Contains an Azide group for CuAAc and SPAAC reactions
Catalog No. 50-003-47596 Supplier Medchemexpress LLC Supplier No. 250MGHY108374
4-Azidobutylamine is a PROTAC linker, characterized by its alkyl chain composition. It is utilized in the synthesis of PROTACs, which are molecules containing two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for a target protein. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins. Additionally, it is a click chemistry reagent containing an Azide group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules, or strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PROTAC linker
  • Click chemistry reagent
  • Contains an Azide group for CuAAc and SPAAC reactions