Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4'-azidocytidine analogue | 876708-01-9 | ≥95.0% | 285.21 | C9H11N5O6 | 10 MG
SDP

Catalog No. 5000297902
Encompass_Preferred

Nucleoside-Analog-2 is a 4'-azidocytidine analogue that inhibits Hepatitis C virus replication and is supplied as a click-chemistry reagent. It contains an azide functional group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC) with suitable partners.

  • 4'-azidocytidine analogue active against HCV replication.
  • Contains an azide group for CuAAC and SPAAC click chemistry.
  • Purity ≥95.0% and molecular weight 285.21 g/mol.
  • Available as solid in multiple mg sizes and as 10 mM solution in DMSO.
  • Powder storage: -20°C for 3 years or 4°C for 2 years; solution storage: -80°C for 2 years.

Catalog No. 50-002-97902 Supplier Medchemexpress LLC Supplier No. HY7765210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nucleoside-Analog-2 is a 4'-azidocytidine analogue that inhibits Hepatitis C virus replication and is supplied as a click-chemistry reagent. It contains an azide functional group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC) with suitable partners.

  • 4'-azidocytidine analogue active against HCV replication.
  • Contains an azide group for CuAAC and SPAAC click chemistry.
  • Purity ≥95.0% and molecular weight 285.21 g/mol.
  • Available as solid in multiple mg sizes and as 10 mM solution in DMSO.
  • Powder storage: -20°C for 3 years or 4°C for 2 years; solution storage: -80°C for 2 years.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.