Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Azidosalicylic Acid 98.0+%, TCI America™
SDP

Catalog No. A22901G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A22901G Supplier TCI America Supplier No. A22901G

Chemical Identifiers

CAS 66761-27-1
Molecular Formula C7H5N3O3
Molecular Weight (g/mol) 179.135
MDL Number MFCD07367665
InChI Key NLPWSMKACWGINL-UHFFFAOYSA-N
Synonym 4-Azido-2-hydroxybenzoic Acid
PubChem CID 3006162
IUPAC Name 4-azido-2-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1N=[N+]=[N-])O)C(=O)O

Specifications

Melting Point 197°C
Quantity 1 g
UN Number 1325
Formula Weight 179.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Azidosalicylic Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.